missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Gut Secretory Cell Epitopes Antibody (FIS 6G5/1) - BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Zebrafish Gut Secretory Cell Epitopes Monoclonal antibody specifically detects Zebrafish Gut Secretory Cell Epitopes in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Zebrafish Gut Secretory Cell Epitopes |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | FIS 6G5/1 |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:200, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
| Formulation | PBS |
| Host Species | Mouse |
| Immunogen | Lysate of zebrafish intestine |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?