missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Gut Secretory Cell Epitopes Antibody (FIS 4B7/2), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50376AF750
This item is not returnable.
View return policy
Description
Zebrafish Gut Secretory Cell Epitopes Monoclonal specifically detects Zebrafish Gut Secretory Cell Epitopes in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Zebrafish Gut Secretory Cell Epitopes | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Lysate of fish intestine | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| FIS 4B7/2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags | |
| Zebrafish | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction