missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Zeba™ Spin Desalting Columns, Plates, and Cartridges, 7K MWCO, 0.5–100 mL

Product Code. 30245279 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 column x 5 mL
10 Columns
2 Plates
25 Columns
4 Plates
5 Columns
5 columns x 1 mL
50 Columns
Format:
96-well Filter Plate
Chromatography Cartridge
Spin Column
Volume (Metric):
0.5 mL
1 mL
10 mL
2 mL
20 mL
5 mL
75 μL
96-well
Unit Size:
Pack of 10
Pack of 2
Pack of 25
Pack of 4
Pack of 5
Pack of 50
This item is not returnable. View return policy

Product Code. 30245279

Brand: Thermo Scientific™ A40002475

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform high-throughput protein recovery (7K MWCO) and sample desalting for mass spectrometry, HPLC, capillary electrophoresis, metabolite screening, and assay development with ready-to-use Thermo Scientific Pierce Zeba desalting chromatography columns, plates, and cartridges.

Perform high-throughput protein recovery and sample desalting for mass spectrometry, HPLC, capillary electrophoresis, metabolite screening, and assay development with ready-to-use Pierce Zeba desalting chromatography columns, plates, and cartridges. Rapidly desalt protein and biological samples for manual or automated liquid chromatography (LC) with ≥95% retention of salts and other small molecular contaminants (<1,000 Da).

Zeba Spin Desalting columns, plates, and chromatography cartridge fittings are convenient, ready-to-use devices compatible with major automated liquid-chromatography systems or for manual syringe processing. The polypropylene columns have a 7K MWCO and contain a proprietary high-performance size-exclusion chromatography resin (Zeba High-Performance Resin) that offers excellent protein desalting and recovery in a centrifuge format. When using Zeba Spin Desalting Columns, even very dilute (25 μg/mL) protein samples can be successfully processed to obtain greater than 95% retention (removal) of salts and other small molecules (<1000 MW), and good recovery of proteins and other macromolecules (>7000 MW).

Similarly, Zeba Desalting plates (7K MWCO) are polypropylene devices that contain Zeba High-Performance Resin and provide excellent protein desalting performance, with high recovery of proteins >7000 Da in a centrifuge format for small sample volumes. The plates enable fast, consistent, well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations. Zeba 96-well spin desalting plates require no resin dispensing or hydration and provide the same high protein recovery as spin columns with at least 95% removal of salts and other small molecules. One plate can process 96 small sample volumes (20 to 100 μL) in just five minutes, offering rapid and exceptional protein desalting.

The Zeba Desalting Chromatography Cartridges contain Zeba High-Performance Resin for desalting or buffer exchange applications. This resin is ideal for removing low molecular weight compounds including salts, fluorescent dyes, biotin, and other small labeling reagents. Protein samples can be processed with unsurpassed protein recovery and 95% retention of salts and other small molecular contaminants (<1,000 Da). Zeba Desalting cartridges provide fast, easy, and reproducible chromatographic separations and can be regenerated for multiple uses.

Benefits of Zeba Spin Desalting columns, plates, and cartridges include:

  • High recovery—low-binding resin maximizes protein recovery
  • Fast—no fraction screening or waiting for protein to emerge by gravity flow
  • Economical—great performance at lower cost than other commercially available cartridges
  • Easy-to-use—no cumbersome column preparation or equilibration
  • Flexible—available in spin columns, filter spin plates and cartridges for a range of needs

Available formats

  • Spin columns—75 μL, 0.5 mL (microcentrifuge), 2 mL, 5 mL, 10 mL columns
  • 96-well spin plates—pre-dispensed 96-well filter plates, compatible with centrifugation for manual or automated purification
  • Chromatography cartridges—compatible with automated liquid chromatography systems and with syringe processing
TRUSTED_SUSTAINABILITY

Specifications

Description Zeba™ Spin Desalting Columns, 7K MWCO, 10 Columns
Formulation 20 mL Zeba Desalting Resin Bed
Product Type Spin Desalting Columns
For Use With (Equipment) Standard Table-top Centrifuge
For Use With (Application) Small molecule removal and buffer exchange
Content And Storage Store at 4°C
Format Spin Column
Purification Target Buffer Exchange, Protein
Column Type Size-exclusion, Proprietary Resin
MWCO 7.0 kDa
Product Line Zeba™
Disposable Single-use
Quantity 10 Columns
Volume (Metric) 20 mL
Sample Volume (Metric) 4 to 10 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.