missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Zeba™ Spin Desalting Columns and Plates, 40K MWCO, 75 μL–10 mL

Product Code. 10470875 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 Plates
25 Columns
4 Plates
5 Columns
50 Columns
Format:
96-well Filter Plate
Spin Column
Volume (Metric):
0.5 mL
10 mL
2 mL
5 mL
75 μL
96-well
Unit Size:
Pack of 2
Pack of 25
Pack of 4
Pack of 5
Pack of 50
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Volume (Metric) unitSize
10470875 25 Columns Spin Column 5 mL Pack of 25
11786436 25 Columns Spin Column 75 μL Pack of 25
10754347 50 Columns Spin Column 75 μL Pack of 50
11796436 25 Columns Spin Column 0.5 mL Pack of 25
10460875 50 Columns Spin Column 0.5 mL Pack of 50
11706446 5 Columns Spin Column 2 mL Pack of 5
10329964 25 Columns Spin Column 2 mL Pack of 25
11716446 5 Columns Spin Column 5 mL Pack of 5
11726446 5 Columns Spin Column 10 mL Pack of 5
10753007 25 Columns Spin Column 10 mL Pack of 25
11582521 2 Plates 96-well Filter Plate 96-well Pack of 2
11592521 4 Plates 96-well Filter Plate 96-well Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 missing translation for 'options'
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10470875 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 87771

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Effectively remove salt and other molecules (40K MWCO) from samples and attain excellent protein recovery with ready-to-use Thermo Scientific Pierce Zeba desalting chromatography columns and plates, which feature a proprietary size-exclusion chromatography resin.

Quickly desalt protein and biological samples for manual or automated liquid chromatography (LC) with Thermo Scientific Zeba Spin and Micro Spin Desalting columns and plates (40K MWCO), which contain Zeba High-Performance Resin, a proprietary size-exclusion chromatography resin that offers excellent protein desalting and recovery in a centrifuge format.

Thermo Scientific Zeba Spin Desalting columns and plates (40K MWCO) are convenient, ready-to-use devices compatible with major automated liquid-chromatography systems or for manual syringe processing. The polypropylene columns have a 40K MWCO and contain a proprietary high-performance size-exclusion chromatography resin (Zeba High-Performance Resin) that offers excellent protein desalting and recovery in a centrifuge format. When using Zeba Micro Spin Desalting Columns, 40K MWCO, 75 μL, even very dilute (25 μg/mL) protein samples can be successfully processed to obtain greater than 95% retention (removal) of salts and other small molecules (<2000 MW), and good recovery of proteins and other macromolecules (>40,000 MW).

Zeba Spin Desalting Plates are polypropylene devices containing a proprietary high-performance size-exclusion resin for effective removal of salts and other small molecules (<2000 daltons) while providing excellent recovery for proteins >40,000 Da. They provide trouble-free desalting and buffer exchange for sample volumes ranging from 20 to 100 μL. This 96-well spin plate format enables more high-throughput processing.

The Zeba 96-well spin desalting plates require no resin dispensing or hydration and provide the same high protein recovery as spin columns with at least 95% removal of salts and other small molecules. One plate can process 96 small sample volumes (20 to 100 μL) in five minutes.

Benefits of Zeba Spin Desalting columns and plates include:
High recovery—low-binding resin maximizes protein recovery
Fast—no fraction screening or waiting for protein to emerge by gravity flow
Economical—great performance at lower cost than other commercially available cartridges
Easy-to-use—no cumbersome column preparation or equilibration
Flexible—available in spin columns and filter spin plates for a range of needs

Available Formats
Spin columns—75 μL, 0.5 mL, 2 mL, 5 mL, 10 mL columns
96-well spin plates—pre-dispensed 96-well filter plates, compatible with centrifugation for manual or automated purification

TRUSTED_SUSTAINABILITY

Especificaciones

Product Type Spin Desalting Column
Content And Storage Upon receipt store at 4°C.
Format Spin Column
Purification Target Buffer Exchange, Protein
Column Type Size-exclusion, Proprietary Resin
MWCO 40 kDa
Product Line Zeba
Quantity 25 Columns
Volume (Metric) 5 mL
Sample Volume (Metric) 300 to 2000 μL
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.