missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZEB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578302
This item is not returnable.
View return policy
Description
Positive Control: MEF, A375, Neuro 2A, C8D30, Raw264.7, C2C12, rat brain Predicted Reactivity: Xenopus laevis (87%), Chicken (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZEB2, initially identified as Smad interacting-protein 1, is normally located in the nucleus and functions as a DNA-binding transcriptional repressor that interacts with activated SMADs. Like the homologous ZEB1, ZEB2 inhibits the transcription of the E-cadherin gene and induces epithelial-mesenchymal transition, a genetic program controlling cell migration during embryonic development and wound healing, in vitro. ZEB2 can also protect cells from DNA damage-induced apoptosis, suggesting that its expression may contribute to tumor progression. Recent evidence has shown that ZEB2 is often observed in the cytoplasm in numerous cancer tissues, indicating that its localization may be regulated in normal and tumor tissues. Mutations in this gene are also associated with Hirschsprung disease/Mowat-Wilson syndrome.
Specifications
| ZEB2 | |
| Polyclonal | |
| Unconjugated | |
| Zeb2 | |
| 9130203F04Rik; D130016B08Rik; HRIHFB2411; HSPC082; KIAA0569; SIP1; SIP-1; SMAD interacting protein 1; smad-interacting protein 1; SMADIP1; Zeb2; ZFHX1B; Zfx1b; Zfxh1b; zinc finger E-box binding homeobox 2; zinc finger E-box-binding homeobox 2; zinc finger homeobox 1b; zinc finger homeobox protein 1b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 24136, 9839 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.42 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O60315, Q9R0G7 | |
| Zeb2 | |
| Recombinant protein encompassing a sequence within the center region of human ZEB2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction