Learn More
ZEB2 Mouse anti-Human, Unconjugated, Clone: 1D3A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67514-1-IG-20UL
This item is not returnable.
View return policy
Description
The ZEB2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZEB2. This antibody recognizes human, mouse, rat antigen. The ZEB2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| ZEB2 | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry | |
| 1D3A2 | |
| Unconjugated | |
| 20 μL | |
| 9839 | |
| IgG1 |