missing translation for 'onlineSavingsMsg'
Learn More

ZEB2 Antibody, HRP, Novus Biologicals™

Product Code. 18426770 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426770 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426770 Supplier Novus Biologicals Supplier No. NBP177179H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZEB2 Polyclonal antibody specifically detects ZEB2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZEB2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias KIAA0569FLJ42816, SIP1HSPC082, Smad-interacting protein 1, SMADIP1ZFHX1BSIP-1, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human ZEB2. The immunogen is located within the last 50 amino acids of ZEB2. Amino Acid Squence: DDSSEDGKMETKSDHEED
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9839
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.