missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZEB2 Antibody (CL13467) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24559MFV450
This item is not returnable.
View return policy
Description
ZEB2 Monoclonal antibody specifically detects ZEB2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| ZEB2 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| KIAA0569FLJ42816, SIP1HSPC082, Smad-interacting protein 1, SMADIP1ZFHX1BSIP-1, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b | |
| This antibody was developed using a recombinant protein derived from O60315, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| 9839 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL13467 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction