missing translation for 'onlineSavingsMsg'
Få mere at vide

ZEB2 Antibody (2H5), Novus Biologicals™

Produktkode 18384199 Shop alle Bio Techne produkter
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mg
Förpackningsstorlek
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity unitSize
18384199 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 18384199 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' H00009839M02

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

Mouse Monoclonal Antibody

ZEB2 Monoclonal antibody specifically detects ZEB2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen ZEB2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2H5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055610
Gene Alias KIAA0569FLJ42816, SIP1HSPC082, Smad-interacting protein 1, SMADIP1ZFHX1BSIP-1, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b
Host Species Mouse
Immunogen ZFHX1B (NP_055610.1, 1115 a.a. ~ 1214 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SITPQGYSDSEERESMPRDGESEKEHEKEGEDGYGKLGRQDGDEEFEEEEEESENKSMDTDPETIRDEEETGDHSMDDSSEDGKMETKSDHEEDNMEDGM
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9839
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.