missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZEB1 Polyclonal Antibody

Description
A suggested positive control is Hela cell lysate. PA5-20979 can be used with blocking peptide PEP-1093.
ZEB1 (Zinc finger E-box-binding homeobox 1) acts as a transcriptional repressor. It inhibits interleukin-2 (IL-2) gene expression. ZEB1 enhances or represses the promoter activity of the ATP1A1 gene depending on the quantity of cDNA and depending on the cell type. It represses E-cadherin promoters and induces an epithelial-mesenchymal transition (EMT) by recruiting SMARCA4/BRG1. ZEB1 also represses BCL6 transcription in the presence of the corepressor CTBP1. It postively regulates neuronal differentiation and represses RCOR1 transcription activation during neurogenesis. Mutations in the gene can result in corneal dystrophy, posterior polymorphous and corneal dystrophy, Fuchs endothelial 6.
Specifications
Specifications
| Antigen | ZEB1 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ZEB1 |
| Gene Accession No. | P37275 |
| Gene Alias | [delta]EF1; 3110032K11Rik; AREB6; BZP; Delta EF1; delta-crystallin enhancer binding factor 1; deltaEF1; DELTA-EF1; FECD6; LOW QUALITY PROTEIN: zinc finger E-box-binding homeobox 1; MEB1; MGC133261; Negative regulator of IL2; Nil2; NIL2A; NIL-2A; NIL-2-A; NIL-2-A zinc finger protein; OTTHUMP00000019405; posterior polymorphous corneal dystrophy 3; PPCD3; RP11-472N13.4; Tcf18; TCF8; TCF-8; Transcription factor 8; transcription factor 8 (represses interleukin 2 expression); Tw; twirler; ZEB; Zeb1; Zfhep; Zfhx1a; Zfx1a; Zfx1ha; zinc finger E-box binding homeobox 1; zinc finger E-box-binding homeobox 1; Zinc finger homeobox protein 1a; zinc finger homeodomain enhancer-binding protein |
| Gene Symbols | ZEB1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?