missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZEB1 Mouse anti-Human, Clone:2A8A6, Abnova™
Mouse monoclonal antibody raised against recombinant human ZEB1.
Brand: Abnova MAB17773.100ug
This item is not returnable.
View return policy
Description
ZEB1 encodes a zinc finger transcription factor that represses T-lymphocyte-specific IL2 gene (MIM 147680) expression by binding to a negative regulatory domain 100 nucleotides 5-prime of the IL2 transcription start site (Williams et al., 1991 [PubMed 1840704]).[supplied by OMIM]Specifications
| ZEB1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry (1:200-1:400) Immunocytochemistry (1:200-1:1000) Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
| ZEB1 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 2A8A6 | |
| zinc finger E-box binding homeobox 1 | |
| In PBS (0.05% sodium azide) | |
| AREB6, BZP, DELTA-EF1, MGC133261, NIL-2-A, NIL-2A, NIL2A, TCF8, ZEB, ZFHEP, ZFHX1A | |
| Recombinant protein corresponding to amino acids 967-1108 of human ZEB1 from E. coli. | |
| 6935 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction