Learn More
ZEB1 Monoclonal Antibody (3G6), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9741-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The monoclonal antibody 3G6 recognizes human, mouse, and rat ZEB1. ZEB1, or zinc finger E-box binding homeobox 1, is also known as Transcription Factor 8 (TCF8), Negative Regulator of IL2, and AREB6. ZEB1 acts as a transcriptional repressor that inhibits interleukin-2 (IL-2) gene expression. ZEB1 also regulates expression of the genes E-cadherin, BCL6, ATP1A1, and RCOR1 through binding to multiple enhancer boxes (E-boxes). ZEB1 promotes epithelial mesenchymal transition (EMT) and invasion of various types of cancers, such as colorectal, breast, pancreatic, lung, and gallbladder. In normal tissues, ZEB1 is expressed in tissues undergoing differentiation such as T cell and skeletal differentiation. Mutations in ZEB1 have been associated with corneal dystrophy. Applications Reported: This 3G6 antibody has been reported for use in western blotting, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This 3G6 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using high pH antigen retrieval and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
ZEB1 (Zinc finger E-box-binding homeobox 1) acts as a transcriptional repressor. It inhibits interleukin-2 (IL-2) gene expression. ZEB1 enhances or represses the promoter activity of the ATP1A1 gene depending on the quantity of cDNA and depending on the cell type. It represses E-cadherin promoters and induces an epithelial-mesenchymal transition (EMT) by recruiting SMARCA4/BRG1. ZEB1 also represses BCL6 transcription in the presence of the corepressor CTBP1. It postively regulates neuronal differentiation and represses RCOR1 transcription activation during neurogenesis. Mutations in the gene can result in corneal dystrophy, posterior polymorphous and corneal dystrophy, Fuchs endothelial 6.Specifications
ZEB1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P37275, Q62947, Q64318 | |
ZEB1 | |
Affinity chromatography | |
RUO | |
21417, 25705, 6935 | |
4° C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
3G6 | |
Unconjugated | |
ZEB1 | |
[delta]EF1; 3110032K11Rik; AREB6; BZP; Delta EF1; delta-crystallin enhancer binding factor 1; deltaEF1; DELTA-EF1; FECD6; LOW QUALITY PROTEIN: zinc finger E-box-binding homeobox 1; MEB1; MGC133261; Negative regulator of IL2; Nil2; NIL2A; NIL-2A; NIL-2-A; NIL-2-A zinc finger protein; OTTHUMP00000019405; posterior polymorphous corneal dystrophy 3; PPCD3; RP11-472N13.4; Tcf18; TCF8; TCF-8; Transcription factor 8; transcription factor 8 (represses interleukin 2 expression); Tw; twirler; ZEB; Zeb1; Zfhep; Zfhx1a; Zfx1a; Zfx1ha; zinc finger E-box binding homeobox 1; zinc finger E-box-binding homeobox 1; Zinc finger homeobox protein 1a; zinc finger homeodomain enhancer-binding protein | |
Mouse | |
25 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
For Research Use Only.