missing translation for 'onlineSavingsMsg'
Learn More

ZEB1 Antibody (2A8A6), PerCP, Novus Biologicals™

Product Code. 18406091 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406091 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406091 Supplier Novus Biologicals Supplier No. NBP223484PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZEB1 Monoclonal antibody specifically detects ZEB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZEB1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2A8A6
Conjugate PerCP
Formulation PBS
Gene Alias AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein
Host Species Mouse
Immunogen Partial recombinant human ZEB1 (between residues 800-1124) expressed in E. coli [Uniprot P37275]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Immunology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 6935
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.