missing translation for 'onlineSavingsMsg'
Learn More

Melford Zeatin

Product Code. 30018968
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018968 1 g 1g
30018966 100 mg 100mg
30018967 250 mg 250mg
30018965 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30018968 Supplier Melford Supplier No. Z114001.0
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Naturally occurring cytokinin growth regulator.

Naturally occurring cytokinin riboside. Main effects in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Chromatographically purified and recrystallized. Plant cell culture tested. Mixed isomers: trans-isomer 97%, cis-isomer >2%.

Specifications

CAS 1637-39-4
Quantity 1 g
Molecular Formula C10H13N5O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.