missing translation for 'onlineSavingsMsg'
Learn More

Zearalenone-4-O-β-D-glucopyranoside, TRC

Product Code. 30457081
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457081 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457081 Supplier Toronto Research Chemicals Supplier No. TRCZ27502525MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zearalenone-4-O-beta-D-glucopyranoside
CAS 105088-14-0
Molecular Formula C24 H32 O10
Quantity 25 mg
Synonym Zearalenone-4-O-beta-D-glucopyranoside
InChI Formula InChI=1S/C24H32O10/c1-13-6-5-9-15(26)8-4-2-3-7-14-10-16(11-17(27)19(14)23(31)32-13)33-24-22(30)21(29)20(28)18(12-25)34-24/h3,7,10-11,13,18,20-22,24-25,27-30H,2,4-6,8-9,12H2,1H3/b7-3+/t13-,18+,20+,21-,22+,24+/m0/s1
SMILES OC1=C2C(=CC(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)=C1)/C=C/CCCC(=O)CCC[C@H](C)OC2=O
IUPAC Name (2E,11S)-15-hydroxy-11-methyl-17-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-12-oxabicyclo[12.4.0]octadeca-1(14),2,15,17-tetraene-7,13-dione
Molecular Weight (g/mol) 480.505
Formula Weight 480.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.