missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ZE 132
Description
ZE 132 binds to programmed cell death-ligand 1 (PD-L1) with high affinity (KD = 19.36 nM); it potently and selectively inhibits the interaction between programmed cell death-1 (PD-1) and PD-L1 (IC50 = 23.49 nM). ZE 132 promotes CD8+ T cell activation and improves the in vitro cytotoxic killing activity of T-cells. ZE 132 treatment can increase Cxcl9 mRNA expression and lower Tgfb1 mRNA expression in CT26 mouse colorectal cancer model. ZE 132 shows antitumor effects, inhibiting growth of CT26 tumors in mice by 64%.
Specifications
Specifications
| Chemical Name or Material | 2-[[[5-Chloro-2-[(5-cyano-3-pyridinyl)methoxy]-4-[[3-(2,3-dihydro-1,4-benzodioxin-6-yl)-2-methylphenyl]methoxy]phenyl]methyl][3-(dimethylamino)propyl]amino]ethanesulfonic acid |
| CAS | 2566710-63-0 |
| Quantity | 2 mg |
| Target | Additional Apoptosis-related Compounds |
| Molecular Formula | C37H41ClN4O7S |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?