missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526721
This item is not returnable.
View return policy
Description
ZDHHC9 Polyclonal Antibody for Western Blot, IHC (P)
The ZDHHC9-GOLGA7 complex is a palmitoyltransferase specific for HRAS and NRAS.
Specifications
| ZDHHC9 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC9 | |
| 6430508G22; 9530098M12Rik; antigen MMSA-1; Asp-His-His-Cys domain containing protein 9; CGI89; CGI-89; CXorf11; DHHC9; DHHC-9; membrane-associated DHHC9 zinc finger protein; MMSA1; MRXSZ; Palmitoyltransferase ZDHHC9; UNQ261/PRO298; ZDHHC10; ZDHHC9; Zinc finger DHHC domain-containing protein 9; zinc finger DHHC-type containing 9; zinc finger protein 379; Zinc finger protein 380; zinc finger, DHHC domain containing 10; zinc finger, DHHC domain containing 9; zinc finger, DHHC-type containing 9; ZNF379; ZNF380 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 208884, 51114 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P59268, Q9Y397 | |
| ZDHHC9 | |
| KLH conjugated synthetic peptide between 335-364 amino acids from the C-terminal region of human ZDHHC9. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction