missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27179-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a four transmembrane protein that is a member of the zinc finger DHHC domain-containing protein family. The encoded protein may function as a palmitoyltransferase. Defects in this gene may be associated with a susceptibility to schizophrenia. Alternate splicing of this gene results in multiple transcript variants. A pseudogene of this gene is found on chromosome 22.Specifications
| ZDHHC8 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC8 | |
| DHHC 8, KIAA1292, ZDHHC8, ZDHHCL1, Zinc finger protein 378, ZNF378 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27801, 29801 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5Y5T5, Q9ULC8 | |
| ZDHHC8 | |
| ZDHHC8 Fusion Protein Ag26028 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction