missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC8 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93430-0.02ml
This item is not returnable.
View return policy
Description
ZDHHC8 Polyclonal antibody specifically detects ZDHHC8 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ZDHHC8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DHHC8, EC 2.3.1, EC 2.3.1.-, KIAA1292DHHC-8, membrane-associated DHHC8 zinc finger protein, probable palmitoyltransferase ZDHHC8, ZDHHCL1, Zinc finger DHHC domain-containing protein 8, Zinc finger protein 378, zinc finger, DHHC-type containing 8, ZNF378zinc finger, DHHC domain like containing 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 240-380 of human ZDHHC8 (NP_037505.1). VEHVLCSPLAPRYVVEPPRLPLAVSLKPPFLRPELLDRAAPLKVKLSDNGLKAGLGRSKSKGSLDRLDEKPLDLGPPLPPKIEAGTFSSDLQTPRPGSAESALSVQRTSPPTPAMYKFRPAFPTGPKVPFCGPGEQVPGPD | |
| 0.02 mL | |
| Neuroscience, Signal Transduction | |
| 29801 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction