missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5116795
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Positive Samples: A-431, PC-3, Mouse lung Immunogen sequence: DFSPPITVIL LIFLCLEGLL FFTFTAVMFG TQIHSICNDE TEIERLKSEK PTWERRLRWE GMKSVFGGPP SLLWMNPFVG FRFRRLPTRP RKGGPEFSV.
ZDHHC7 is a protein coding gene. Palmitoyltransferase with broad specificity. Palmitoylates JAM3. Palmitoylates SNAP25 and DLG4/PSD95. Palmitoylates sex steroid hormone receptors, including ESR1, PGR and AR, thereby regulating their targeting to the plasma membrane and their function in rapid intracellular signaling upon binding of sex hormones. May play a role in follicle stimulation hormone (FSH) activation of testicular Sertoli cells.
Spezifikation
| ZDHHC7 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC7 | |
| AL024087; DHHC7; DHHC-7; GABA-A receptor-associated membrane protein 2; Gramp2; membrane-associated DHHC7 zinc finger protein; palmitoyltransferase ZDHHC7; putative zinc finger protein SERZ-1; sertoli cell gene with a zinc finger domain protein; Sertoli cell gene with zinc finger domainβ Serz; SERZ1; Serz-1; SERZ-B; Zdhhc7; Zinc finger DHHC domain-containing protein 7; zinc finger DHHC-type containing 7; zinc finger protein 370; zinc finger, DHHC domain containing 7; zinc finger, DHHC-type containing 7; ZNF370 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 102193, 55625 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 4.45 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q91WU6, Q9NXF8 | |
| ZDHHC7 | |
| A synthetic peptide corresponding to a sequence within amino acids 210 to the C-terminus of human ZDHHC7 (NP_0602102). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur