missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32486
This item is not returnable.
View return policy
Description
ZDHHC7 Polyclonal specifically detects ZDHHC7 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ZDHHC7 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| EC 2.3.1, EC 2.3.1.-, FLJ10792, FLJ20279, palmitoyltransferase ZDHHC7, Sertoli cell gene with zinc finger domain- and #946, SERZ1, SERZ-B, Zinc finger DHHC domain-containing protein 7, Zinc finger protein 370, zinc finger, DHHC domain containing 7, zinc finger, DHHC-type containing 7, ZNF370DHHC-7 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55625 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ZDHHC7 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MQPSGHRLRDVEHHPLLAENDNYDSSSSS | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction