missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £364.00
Specifications
| Antigen | ZDHHC6 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ZDHHC6 Polyclonal antibody specifically detects ZDHHC6 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ZDHHC6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 64429 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DHHC-6, EC 2.3.1, EC 2.3.1.-, FLJ21952, Transmembrane protein H4, Zinc finger DHHC domain-containing protein 6, Zinc finger protein 376, zinc finger, DHHC domain containing 6, zinc finger, DHHC-type containing 6, ZNF376probable palmitoyltransferase ZDHHC6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 279-413 of human ZDHHC6 (NP_071939.1). SGVPEGDGLEWPVREGCHQYSLTIEQLKQKADKRVRSVRYKVIEDYSGACCPLNKGIKTFFTSPCTEEPRIQLQKGEFILATRGLRYWLYGDKILDDSFIEGVSRIRGWFPRKCVEKCPCDAETDQAPEGEKKNR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title