missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566681
This item is not returnable.
View return policy
Description
Immunogen sequence: VYEFDEVMFPK NVRCSTCDLR KPARSK Highest antigen sequence identity to the following orthologs - mouse 85%, rat 96%.
ZDHHC4 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; protein-cysteine S-palmitoyltransferase activity.
Specifications
| ZDHHC4 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC4 | |
| 1810021D01Rik; 2900029I10Rik; DC1; DHHC-4; membrane-associated DHHC4 zinc finger protein; probable palmitoyltransferase ZDHHC4; UNQ5787/PRO19576; Zdhhc4; Zinc finger DHHC domain-containing protein 4; zinc finger DHHC-type containing 4; zinc finger protein 374; zinc finger, DHHC domain containing 4; zinc finger, DHHC-type containing 4; ZNF374 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55146 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NPG8 | |
| ZDHHC4 | |
| Recombinant Human ZDHHC4. Recombinant protein control fragment (Product #RP-107330). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction