missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572909
This item is not returnable.
View return policy
Description
ZDHHC23 Polyclonal Antibody for Western Blot, IHC (P), IP
Palmitoyltransferase that mediates palmitoylation of KCNMA1, regulating localization of KCNMA1 to the plasma membrane. May be involved in NOS1 regulation and targeting to the synaptic membrane.
Specifications
| ZDHHC23 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC23 | |
| DHHC-23; DHHC-containing protein 11; Gm1751; Gm779; NIDD; nNOS-intereacting DHHC-containing Dem protein-L; palmitoyltransferase ZDHHC23; probable palmitoyltransferase ZDHHC23; Zdhhc23; Zinc finger DHHC domain-containing protein 23; zinc finger DHHC-type containing 23; zinc finger, DHHC domain containing 23 | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Conc. Not Determined | |
| Whole serum with 0.02% sodium azide | |
| Q8IYP9 | |
| ZDHHC23 | |
| Full-length recombinant human NIDD protein. | |
| RUO | |
| 254887 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction