missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116793
This item is not returnable.
View return policy
Description
Positive Samples: BxPC-3, Rat lung Immunogen sequence: QVFGDEKKYW LLPIFSSLGD GCSFPTCLVN QDPEQASTPA GLNSTAKNLE NHQFPAKPLR ESQSHLLTDS QSWTESSINP GKCKAGMSNP ALTMENET.
Palmitoyltransferase that catalyzes the addition of palmitate onto various protein substrates and is involved in a variety of cellular processes (PubMed:18508921, PubMed:18296695, PubMed:19144824, PubMed:21343290, PubMed:22034844, PubMed:23793055). In the nervous system, plays a role in long term synaptic potentiation by palmitoylating AKAP5 through which it regulates protein trafficking from the dendritic recycling endosomes to the plasma membrane and controls both structural and functional plasticity at excitatory synapses. In dendrites, mediates the palmitoylation of DLG4 when synaptic activity decreases and induces synaptic clustering of DLG4 and associated AMPA-type glutamate receptors. Also mediates the de novo and turnover palmitoylation of RGS7BP, a shuttle for Gi/o-specific GTPase-activating proteins/GAPs, promoting its localization to the plasma membrane in response to the activation of G protein-coupled receptors. Through the localization of these GTPase-activating proteins/GAPs, it also probably plays a role in G protein-coupled receptors signaling in neurons. Also probably plays a role in cell adhesion by palmitoylating CD9 and CD151 to regulate their expression and function (PubMed:18508921). Palmitoylates the endoplasmic reticulum protein CKAP4 and regulates its localization to the plasma membrane (PubMed:18296695, PubMed:19144824). Could also palmitoylate LCK and regulate its localization to the plasma membrane (PubMed:22034844). [UniProt].
Specifications
| ZDHHC2 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC2 | |
| 5730415P04Rik; 6430583A19Rik; DHHC2; DHHC-2; membrane-associated DHHC2 zinc finger protein; palmitoyltransferase ZDHHC2; Ream; rec; Reduced expression associated with metastasis protein; reduced expression in cancer protein; small rec; srec; testis tissue sperm-binding protein Li 56e; Zdhhc2; Zinc finger DHHC domain-containing protein 2; zinc finger DHHC-type containing 2; zinc finger protein 372; zinc finger, DHHC domain containing 2; zinc finger, DHHC-type containing 2; ZNF372 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 246326, 51201 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.93 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9JKR5, Q9UIJ5 | |
| ZDHHC2 | |
| A synthetic peptide corresponding to a sequence within amino acids 270-367 of human ZDHHC2 (NP_0574371). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction