missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575752
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZDHHC1 (zinc finger, DHHC-type containing 1), also known as ZNF377 (zinc finger protein 377) or C16orf1, is a 485 amino acid multi-pass membrane protein that contains one DHHC-type zinc finger. Expressed in fetal heart, lung and kidney and also present in adult lung and pancreas, ZDHHC1 functions as a palmitoyltransferase that catalyzes the conversion of palmitoyl-CoA and protein-cysteine to S-palmitoyl protein and CoA. Like ZDHHC1, ZDHHC5, ZDHHC6, ZDHHC7 and ZDHHC18 each contain one DHHC-type zinc finger through which they convey palmitoyltransferase activity against a broad range of substrates, including H-Ras, SNAP 25 and GABAA R proteins.
Specifications
| ZDHHC18 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC18 | |
| DHHC18; DHHC-18; membrane-associated DHHC18 zinc finger protein; palmitoyltransferase ZDHHC18; Zdhhc18; Zinc finger DHHC domain-containing protein 18; zinc finger DHHC-type containing 18; zinc finger, DHHC domain containing 18; zinc finger, DHHC-type containing 18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84243 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NUE0 | |
| ZDHHC18 | |
| Synthetic peptide corresponding to residues in Human ZDHHC18. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction