missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118804
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZDHHC17.
Palmitoyltransferase specific for a subset of neuronal proteins, including SNAP25, DLG4/PSD95, GAD2, SYT1 and HD. May be involved in the sorting or targeting of critical proteins involved in the initiating events of endocytosis at the plasma membrane. Has transforming activity. Mediates Mg(2+) transport.
Specifications
| ZDHHC17 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC17 | |
| A230053P19Rik; BB187739; D130071N24Rik; DHHC-17; Hip14; HIP-14; HIP3; HIP-3; HSPC294; huntingtin interacting protein 14; huntingtin interacting protein 3; Huntingtin interacting protein H; huntingtin yeast partner H; Huntingtin-interacting protein 14; huntingtin-interacting protein 3; Huntingtin-interacting protein H; HYPH; Kiaa0946; membrane-associated DHHC17 zinc finger protein; palmito; palmitoyltransferase ZDHHC17; Putative MAPK-activating protein PM11; putative NF-kappa-B-activating protein 205; ZDHHC17; Zinc finger DHHC domain-containing protein 17; zinc finger DHHC-type containing 17; zinc finger, DHHC domain containing 17; zinc finger, DHHC-type containing 17 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23390, 320150, 366889 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| E9PTT0, Q80TN5, Q8IUH5 | |
| ZDHHC17 | |
| A synthesized peptide derived from human ZDHHC17(Accession Q8IUH5), corresponding to amino acid residues K257-L307. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction