missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £443.00
Specifications
| Antigen | ZDHHC17 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18476810
|
Novus Biologicals
NBP1-81748-25ul |
25 μL |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18244397
|
Novus Biologicals
NBP1-81748 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZDHHC17 Polyclonal specifically detects ZDHHC17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ZDHHC17 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 23390 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RKEGFNTKMADGPDEYDTEAGCVPLLHPEEIKPQSHYNHGYGEPLGRKTHIDDYSTWDIVKATQYGIYERCRELVEAGYDVR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| DHHC-17, EC 2.3.1, EC 2.3.1.-, HIP14HIP-14, huntingtin interacting protein 14, huntingtin interacting protein 3, Huntingtin interacting protein H, Huntingtin yeast partner H, Huntingtin-interacting protein 14, Huntingtin-interacting protein 3, Huntingtin-interacting protein H, HYPHHIP-3, KIAA0946HIP3, palmitoyltransferase ZDHHC17, Putative MAPK-activating protein PM11, Putative NF-kappa-B-activating protein 205, Zinc finger DHHC domain-containing protein 17, zinc finger, DHHC domain containing 17, zinc finger, DHHC-type containing 17 | |
| ZDHHC17 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title