missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ZDHHC16 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZDHHC16 Polyclonal specifically detects ZDHHC16 in Human samples. It is validated for Western Blot.Specifications
| ZDHHC16 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| DHHC domain containing 16, EC 2.3.1, EC 2.3.1.-, zinc finger, DHHC-type containing 16 | |
| ZDHHC16 | |
| IgG | |
| 44 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q969W1 | |
| 84287 | |
| Synthetic peptides corresponding to ZDHHC16(zinc finger, DHHC-type containing 16) The peptide sequence was selected from the C terminal of ZDHHC16. Peptide sequence VLISRGETSIERHINKKERRRLQAKGRVFRNPYNYGCLDNWKVFLGVDTG. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title