missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539327
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the DHHC palmitoyltransferase family. Mutations in this gene are associated with mental retardatio X-linked type 91 (MRX91). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2009].
Specifications
| ZDHHC15 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC15 | |
| ZDHHC15 | |
| A synthetic peptide derived from the C-terminal region of human ZDHHC15 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96MV8, Q8BGJ0, Q2TGJ4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 108672, 158866, 317235 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction