missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101738
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZDHHC15.
The protein encoded by this gene belongs to the DHHC palmitoyltransferase family. Mutations in this gene are associated with mental retardatio X-linked type 91 (MRX91). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2009].
Specifications
| ZDHHC15 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC15 | |
| 6030457O13Rik; DHHC-15; membrane-associated DHHC15 zinc finger protein; MRX91; Palmitoyltransferase ZDHHC15; UNQ1969/PRO4501; Zdhhc15; Zinc finger DHHC domain-containing protein 15; zinc finger DHHC-type containing 15; zinc finger, DHHC domain containing 15; zinc finger, DHHC-type containing 15 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 108672, 158866 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BGJ0, Q96MV8 | |
| ZDHHC15 | |
| A synthesized peptide derived from human ZDHHC15(Accession Q96MV8), corresponding to amino acid residues H293-T337. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction