missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582559
This item is not returnable.
View return policy
Description
Immunogen sequence: IAYLISKGQS VNMTDVNGQT PLMLSAHKVI GPEPTGFLLK FNPSLNVVDK IHQNTPLHWA VAAGNVNAVD KLLEAGSSLD IQNVKGETPL DMALQN Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 93%.
Palmitoyltransferase for HTT and GAD2. May play a role in Mg(2+) transport. [UniProt].
Specifications
| ZDHHC13 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC13 | |
| 2410004E01Rik; C530010M18; DHHC-13; fi22e09; FLJ10852; FLJ10941; Hip14l; HIP14-like; HIP14-related protein; HIP3RP; huntingtin interacting protein HIP3RP; huntingtin-interacting protein 14-related protein; Huntingtin-interacting protein HIP3RP; hypothetical protein LOC494107; kojak; MGC64994; Palmitoyltransferase ZDHHC13; probable palmitoyltransferase ZDHHC13; putative MAPK-activating protein PM03; Putative NF-kappa-B-activating protein 209; skc4; wu:fb06e01; wu:fc39g10; wu:fi22e09; Zdhhc13; zgc:101690; Zinc finger DHHC domain-containing protein 13; zinc finger DHHC-type containing 13; zinc finger DHHC-type palmitoyltransferase 13; zinc finger, DHHC domain containing 13; zinc finger, DHHC-type containing 13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54503 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IUH4 | |
| ZDHHC13 | |
| Recombinant protein corresponding to Human ZDHHC13. Recombinant protein control fragment (Product #RP-91300). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction