missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC13 Monoclonal Antibody (2A4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522031
This item is not returnable.
View return policy
Description
Peptide Sequence: NGQTPLMLSA HKVIGPEPTG FLLKFNPSLN VVDKIHQNTP LHWAVAAGNV NAVDKLLEAG SSLDIQNVKG ETPLDMALQN KNQLIIHMLK TEAKMRANQK.
Palmitoyltransferase for HTT and GAD2. May play a role in Mg(2+) transport. [UniProt].
Specifications
| ZDHHC13 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8IUH4 | |
| ZDHHC13 | |
| ZDHHC13 (NP_061901.2, 181 a.a. ~ 280 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2A4 | |
| Unconjugated | |
| ZDHHC13 | |
| 2410004E01Rik; C530010M18; DHHC-13; fi22e09; FLJ10852; FLJ10941; Hip14l; HIP14-like; HIP14-related protein; HIP3RP; huntingtin interacting protein HIP3RP; huntingtin-interacting protein 14-related protein; Huntingtin-interacting protein HIP3RP; hypothetical protein LOC494107; kojak; MGC64994; Palmitoyltransferase ZDHHC13; probable palmitoyltransferase ZDHHC13; putative MAPK-activating protein PM03; Putative NF-kappa-B-activating protein 209; skc4; wu:fb06e01; wu:fc39g10; wu:fi22e09; Zdhhc13; zgc:101690; Zinc finger DHHC domain-containing protein 13; zinc finger DHHC-type containing 13; zinc finger DHHC-type palmitoyltransferase 13; zinc finger, DHHC domain containing 13; zinc finger, DHHC-type containing 13 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54503 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction