missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570499
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 79%; Rat: 93%.
ZDHHC12 gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein palmitoylation; protein targeting to membrane.
Specifications
| ZDHHC12 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC12 | |
| 1190004A01Rik; DHHC-12; membrane-associated DHHC12 zinc finger protein; probable palmitoyltransferase ZDHHC12; PSEC0008; ZDHHC12; Zinc finger DHHC domain-containing protein 12; zinc finger DHHC-type containing 12; zinc finger DHHC-type palmitoyltransferase 12; zinc finger protein 400; zinc finger, DHHC domain containing 12; zinc finger, DHHC-type containing 12; ZNF400 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84885 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96GR4 | |
| ZDHHC12 | |
| synthetic peptide directed towards the N-terminal region of human ZDHHC12. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction