missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZDHHC11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13462617
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13462617 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13462617 Supplier Invitrogen™ Supplier No. PA528503

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116, MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZDHHC11 gene ontology annotations related to this gene include peptidyl-L-cysteine S-palmitoylation; protein targeting to membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZDHHC11
Gene Accession No. Q9H8X9
Gene Alias 4933421L13Rik; DHHC-11; DHHC-containing protein 10; LOC100057809; membrane-associated DHHC11 zinc finger protein; probable palmitoyltransferase ZDHHC11; Zdhhc11; Zdhhc20; Zinc finger DHHC domain-containing protein 11; zinc finger DHHC-type containing 11; zinc finger protein 399; zinc finger, DHHC domain containing 11; zinc finger, DHHC domain containing 20; zinc finger, DHHC-type containing 11; Znf399
Gene Symbols ZDHHC11
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZDHHC11. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79844
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.