missing translation for 'onlineSavingsMsg'
Learn More

ZCWPW2 Antibody (OTI10A12) [Alexa Fluor™ 700], Novus Biologicals™

Product Code. 30687816 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687816 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687816 Supplier Novus Biologicals Supplier No. NBP344988AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZCWPW2 Monoclonal antibody specifically detects ZCWPW2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCWPW2
Applications Western Blot
Classification Monoclonal
Clone OTI10A12
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias ZCW2, zinc finger CW-type PWWP domain protein 2, zinc finger, CW type with PWWP domain 2
Host Species Mouse
Immunogen Full length human recombinant protein of human ZCWPW2 (NP_001035522) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152098
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.