missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZCRB1 Polyclonal Antibody

Description
A suggested positive control is Raji cell lysate. PA5-20772 can be used with blocking peptide PEP-0886.
ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.
Specifications
Specifications
| Antigen | ZCRB1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Zcrb1 |
| Gene Accession No. | Q499V6, Q8TBF4, Q9CZ96 |
| Gene Alias | 2700088M22Rik; MADP1; Madp-1; MADP-1 protein; MADP-1 protein-like; MGC26805; RBM36; RGD1309851; SNRNP31; U11/U12 small nuclear ribonucleoprotein 31 kDa protein; U11/U12 snRNP 31 kDa protein; U11/U12 snRNP 31K; U11/U12-31K; ZCCHC19; Zcrb1; zinc finger CCHC-type and RNA binding motif 1; zinc finger CCHC-type and RNA binding motif containing 1; zinc finger CCHC-type and RNA-binding motif-containing protein 1 |
| Gene Symbols | Zcrb1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?