missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCRB1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76319PCP
This item is not returnable.
View return policy
Description
ZCRB1 Polyclonal antibody specifically detects ZCRB1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| ZCRB1 | |
| Polyclonal | |
| Western Blot, ELISA | |
| MADP-1, MADP1U11/U12 snRNP 31 kDa protein, MGC26805, RBM36, U11/U12 small nuclear ribonucleoprotein 31 kDa protein, U11/U12 snRNP 31K, U11/U12-31K, ZCCHC19, zinc finger CCHC-type and RNA binding motif 1, zinc finger CCHC-type and RNA-binding motif-containing protein 1 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZCRB1. The immunogen is located within the last 50 amino acids of ZCRB1. Amino Acid Squence: TSDDSRRPRIKKSTYFS | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 85437 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction