missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCCHC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25196-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| ZCCHC6 | |
| Polyclonal | |
| Unconjugated | |
| ZCCHC6 | |
| HS2, KIAA1711, PAPD6, Terminal uridylyltransferase 7, TUT7, TUTase 7, ZCCHC6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 214290, 501515, 79670 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5BLK4, Q5VYS8 | |
| Zcchc6 | |
| ZCCHC6 Fusion Protein Ag18240 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction