Learn More
ZCCHC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25196-1-AP-20UL
 View more versions of this product
Product Code. 16863755
  • £135.00 / 20µL
Description
The function of this protein remains unknown.
Specifications
Specifications
ZCCHC6
Polyclonal
Unconjugated
ZCCHC6
HS2, KIAA1711, PAPD6, Terminal uridylyltransferase 7, TUT7, TUTase 7, ZCCHC6
Rabbit
Antigen Affinity Chromatography
RUO
214290, 501515, 79670
-20°C
Liquid
Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5BLK4, Q5VYS8
Zcchc6
ZCCHC6 Fusion Protein Ag18240
20 μL
Primary
Mouse, Human, Rat