missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZCCHC3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA543623

Product Code. 15576142

  • £442.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Peptide sequence: EKKGEFREPR PPRREEESGG GGGSAGLGGP AGLAAPDLGD FPPAGRGDPK Sequence homology: Cow: 85%; Human: 100%; Pig: 85%.

Nucleic acid-binding protein involved in innate immune response to DNA and RNA viruses (PubMed:30193849, PubMed:30135424). Binds DNA and RNA in the cytoplasm and acts by promoting recognition of viral nucleic acids by virus sensors, such as DDX58/RIG-I, IFIH1/MDA5 and CGAS (PubMed:30193849, PubMed:30135424). Acts as a co-sensor for recognition of double-stranded DNA (dsDNA) by cGAS in the cytoplasm, thereby playing a role in innate immune response to cytosolic dsDNA and DNA virus (PubMed:30135424). Binds dsDNA and probably acts by promoting sensing of dsDNA by CGAS, leading to enhance CGAS oligomerization and activation (PubMed:30135424). Promotes sensing of viral RNA by RIG-I-like receptors proteins DDX58/RIG-I and IFIH1/MDA5 via two mechanisms: binds double-stranded RNA (dsRNA), enhancing the binding of DDX58/RIG-I and IFIH1/MDA5 to dsRNA and promotes 'Lys-63'-linked ubiquitination and subsequent activation of DDX58/RIG-I and IFIH1/MDA5 (PubMed:30193849). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZCCHC3
Polyclonal
Unconjugated
ZCCHC3
C20orf99; ZCCHC3; Zinc finger CCHC domain-containing protein 3; zinc finger CCHC-type containing 3; zinc finger, CCHC domain containing 3
Rabbit
Affinity Chromatography
RUO
85364
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9NUD5
ZCCHC3
Synthetic peptide directed towards the N-terminal of human ZCCHC3 (aa 49-98).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.