missing translation for 'onlineSavingsMsg'
Learn More

ZCCHC3 Antibody, Novus Biologicals™

Product Code. 18414689 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414689 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18414689 Leverantör Novus Biologicals Leverantörsnummer H00085364B01P

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Polyclonal Antibody

ZCCHC3 Polyclonal antibody specifically detects ZCCHC3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen ZCCHC3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_149080
Gene Alias C20orf99, chromosome 20 open reading frame 99, dJ1103G7.7, MGC104290, zinc finger CCHC domain-containing protein 3, zinc finger, CCHC domain containing 3
Host Species Mouse
Immunogen ZCCHC3 (NP_149080, 1 a.a. - 403 a.a.) full-length human protein. MATGGGAEEERKRGRPQLLPPARPAARGEEADGGREKMGWAQVVKNLAEKKGEFREPRPPRREEESGGGGGSAGLGGPAGLAAPDLGDFPPAGRGDPKGRRRDPAGEAVDPRKKKGAAEAGRRKKAEAAAAAMATPARPGEAEDAAERPLQDEPAAAAGPGKGRFLVRICFQGDEGACPTRDFVVGALILRSIGMDPSDIYAVIQIPGSREFDVSFRSAEKLALFLRVYEEKREQEDCWENFVVLGRSKSSLKTLFILFRNETVDVEDIVTWLKRHCDVLAVPVKVTDRFGIWTGEYKCEIELRQGEGGVRHLPGAFFLGAERGYSWYKGQPKTCFKCGSRTHMSGSCTQDRCFRCGEEGHLSPYCRKGIVCNLCGKRGHAFAQCPKAVHNSVAAQLTGVAGH
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 85364
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.