missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCCHC3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | ZCCHC3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663952
|
Novus Biologicals
NBP2-94056-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18626412
|
Novus Biologicals
NBP2-94056-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZCCHC3 Polyclonal antibody specifically detects ZCCHC3 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| ZCCHC3 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 85364 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| C20orf99, chromosome 20 open reading frame 99, dJ1103G7.7, MGC104290, zinc finger CCHC domain-containing protein 3, zinc finger, CCHC domain containing 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 215-335 of human ZCCHC3 (NP_149080.2). SFRSAEKLALFLRVYEEKREQEDCWENFVVLGRSKSSLKTLFILFRNETVDVEDIVTWLKRHCDVLAVPVKVTDRFGIWTGEYKCEIELRQGEGGVRHLPGAFFLGAERGYSWYKGQPKTC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title