missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZCCHC17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584169
This item is not returnable.
View return policy
Description
Immunogen sequence: MNSGRPETME NLPALYTIFQ GEVAMVTDYG AFIKIPGCRK QGLVHRTHMS SCRVDKPSEI VDVGDKVWVK LIGREMK Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
ZCCHC17 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytosolic large ribosomal subunit; nucleolus; nucleic acid binding; zinc ion binding.
Specifications
| ZCCHC17 | |
| Polyclonal | |
| Unconjugated | |
| ZCCHC17 | |
| 2810055E05Rik; hpNO40s; HSPC243; HSPC251; LDC4; nucleolar protein 40; nucleolar protein of 40 kDa; Pnn-interacting nucleolar protein; pNO40; PS1D; PS1D protein; putative S1 RNA binding domain protein; putative S1 RNA-binding domain protein; RGD1565267; Zcchc17; Zinc finger CCHC domain-containing protein 17; zinc finger CCHC-type containing 17; zinc finger, CCHC domain containing 17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51538 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NP64 | |
| ZCCHC17 | |
| Recombinant protein corresponding to Human ZCCHC17. Recombinant protein control fragment (Product #RP-106491). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction