missing translation for 'onlineSavingsMsg'
Learn More

ZCCHC17 Antibody (OTI1F11) [DyLight 488], Novus Biologicals™

Product Code. 30688674 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30688674 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688674 Supplier Novus Biologicals Supplier No. NBP344966G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZCCHC17 Monoclonal antibody specifically detects ZCCHC17 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC17
Applications Western Blot
Classification Monoclonal
Clone OTI1F11
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias HSPC251, nucleolar protein of 40 kDa, Pnn-interacting nucleolar protein, pNO40nucleolar protein 40, PS1D protein, PS1DRP11-266K22.1, putative S1 RNA binding domain protein, Putative S1 RNA-binding domain protein, Zinc finger CCHC domain-containing protein 17, zinc finger, CCHC domain containing 17
Host Species Mouse
Immunogen Full length human recombinant protein of human ZCCHC17 (NP_057589) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51538
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.