missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ZCCHC11/TUT4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29254
This item is not returnable.
View return policy
Description
ZCCHC11/TUT4 Polyclonal antibody specifically detects ZCCHC11/TUT4 in Human,Mouse samples. It is validated for Immunohistochemistry
Specifications
| ZCCHC11/TUT4 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry | |
| Unconjugated | |
| DKFZp779C1943, EC 2.7.7.52, KIAA0191FLJ42878, PAP associated domain containing 3, PAPD3, terminal uridylyltransferase 4, TUT4, TUTase 4, Zinc finger CCHC domain-containing protein 11, zinc finger, CCHC domain containing 11 | |
| Between 250 and 300 | |
| 25 μg | |
| Cell Cycle and Replication | |
| 23318 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction