missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCCHC10 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94623-0.1ml
This item is not returnable.
View return policy
Description
ZCCHC10 Polyclonal antibody specifically detects ZCCHC10 in Mouse samples. It is validated for Western Blot
Specifications
| ZCCHC10 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ20094, zinc finger CCHC domain-containing protein 10, zinc finger, CCHC domain containing 10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human ZCCHC10 (NP_001287745.1). MATPMHRLIARRQAFDTELQPVKTFWILIQPSIVISEANKQHVRCQKCLEFGHWTYECTGKRKYLHRPSRTAELKKALKEKENRLLLQQSIGETNVERKA | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54819 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction