missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H8 Antibody (OTI4H5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46397
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Immunohistochemistry
ZC3H8 Monoclonal specifically detects ZC3H8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ZC3H8 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:150 | |
| Fliz1, ZC3HDC8, zinc finger CCCH domain-containing protein 8, zinc finger CCCH type domain containing 8, zinc finger CCCH-type containing 8, zinc finger CCCH-type domain containing 8 | |
| Mouse | |
| 33.4 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| OTI4H5 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| ZC3H8 | |
| Full length human recombinant protein of human ZC3H8 (NP_115883) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| 84524 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction