missing translation for 'onlineSavingsMsg'
Learn More

ZC3H8 Antibody (OTI4H5), DyLight 594, Novus Biologicals™

Product Code. 18429455 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429455 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429455 Supplier Novus Biologicals Supplier No. NBP274908DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZC3H8 Monoclonal antibody specifically detects ZC3H8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H8
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI4H5
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Fliz1, ZC3HDC8, zinc finger CCCH domain-containing protein 8, zinc finger CCCH type domain containing 8, zinc finger CCCH-type containing 8, zinc finger CCCH-type domain containing 8
Host Species Mouse
Immunogen Full length recombinant protein of human ZC3H8 (NP_115883) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84524
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.