missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H8 Antibody (OTI4H5), CoraFluor™ 1, Novus Biologicals™
Description
ZC3H8 Monoclonal antibody specifically detects ZC3H8 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ZC3H8 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OTI4H5 |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | Fliz1, ZC3HDC8, zinc finger CCCH domain-containing protein 8, zinc finger CCCH type domain containing 8, zinc finger CCCH-type containing 8, zinc finger CCCH-type domain containing 8 |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human ZC3H8 (NP_115883) produced in E.coli. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?